ContactUsLinkComponent
MitoROS™ 580 Kit for Detecting Reactive Oxygen Species (ROS) in Mitochondria
MitoROS™ 580 Kit for Detecting Reactive Oxygen Species (ROS) in Mitochondria
Catalog # AATB16052
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
MitoROS™ 580 Kit for Detecting Reactive Oxygen Species (ROS) in Mitochondria
Catalog # AATB16052
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    500 Tests
  • Description:
    Superoxide-sensitive dye that is localized in mitochondria upon loading into live cells
Specifications
About this item
Oxidation of MitoROS™ 580 by superoxide generates red fluorescence.
  • Optimized for mitochondria
  • Live cell reagent
MitoROS™ 580 reagent permeates live cells where it selectively targets mitochondria. It is rapidly oxidized by superoxide. It is less likely to be oxidized by other reactive oxygen species (ROS) and reactive nitrogen species (RNS). The oxidized product is highly fluorescent in cells.
Packaging: Plastic vial.
Caution: For research use only.