ReadiLink™ KLH Conjugation Kit (2 Reactions)
Catalog # AATB5502
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:2 Tests
- Description:ReadiLink™ KLH conjugation kit
Specifications
About this item
Keyhole Limpet Hemocyanin (KLH) is one of the most commonly used carriers in the conjugation of peptides for antibody production.
- Excellent protein recovery
- Stable conjugation
- Easy and simple protocol
Caution: For research use only.