ContactUsLinkComponent
ReadiLink™ Rapid mFluor™ Blue 570 Antibody Labelling Kit 'Microscale Optimised for Labelling 50 µg Antibody Per Reaction' (2 Labellings)
ReadiLink™ Rapid mFluor™ Blue 570 Antibody Labelling Kit 'Microscale Optimised for Labelling 50 µg Antibody Per Reaction' (2 Labellings)
Katalognr. AATB1120
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
ReadiLink™ Rapid mFluor™ Blue 570 Antibody Labelling Kit 'Microscale Optimised for Labelling 50 µg Antibody Per Reaction' (2 Labellings)
Katalognr. AATB1120
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    2 Tests
  • Description:
    ReadiLink™ rapid mFluor™ blue 570 antibody labeling kit 'Microscale optimized for labeling 50 µg antibody per reaction'
Specifications
About this item
AAT Bioquest's mFluor™ dyes are developed for flow cytometry-focused applications.
  • Blue-laser excitable
  • Robust and simple antibody labelling protocol
  • No column purification required
Caution: For research use only.