ContactUsLinkComponent
ReadiLink™ Rapid mFluor™ Violet 510 Antibody Labelling Kit 'Microscale Optimised for Labelling 50 µg Antibody Per Reaction' (2 Labellings)
ReadiLink™ Rapid mFluor™ Violet 510 Antibody Labelling Kit 'Microscale Optimised for Labelling 50 µg Antibody Per Reaction' (2 Labellings)
  AATB1110
 :  
undefined
ReadiLink™ Rapid mFluor™ Violet 510 Antibody Labelling Kit 'Microscale Optimised for Labelling 50 µg Antibody Per Reaction' (2 Labellings)
  AATB1110
 :  
  • Pk:
    2 Tests
  • Description:
    ReadiLink™ rapid mFluor™ violet 510 antibody labeling kit microscale optimised for labeling 50 µg antibody per reaction
AAT Bioquest's mFluor™ dyes are developed for flow cytometry-focused applications.
  • Violet-laser excitable
  • Robust and simple antibody labelling protocol
  • No column purification required
 : For research use only.