ReadiLink™ Rapid mFluor™ Violet 450 Antibody Labelling Kit 'Microscale Optimised for Labelling 50 µg Antibody Per Reaction' (2 Labellings)
Katalognr. AATB1100
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- VE:2 Tests
- Bezeichnung:ReadiLink™ rapid mFluor™ violet 450 antibody labeling kit 'Microscale optimized for labeling 50 µg antibody per reaction'
Specifications
About this item
AAT Bioquest's mFluor™ dyes are developed for flow cytometry-focused applications.
- Suitable for Pacific Blue™ channel
- robust and simple antibody labelling protocol
- no column purification required
Caution: For research use only.