Order Entry
Switzerland
ContactUsLinkComponent
ReadiLink™ Rapid Cy7 Antibody Labelling Kit 'Microscale Optimised for Labelling 50 µg Antibody Per Reaction' (2 Labellings)
ReadiLink™ Rapid Cy7 Antibody Labelling Kit 'Microscale Optimised for Labelling 50 µg Antibody Per Reaction' (2 Labellings)
Catalog # AATB1294
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
ReadiLink™ Rapid Cy7 Antibody Labelling Kit 'Microscale Optimised for Labelling 50 µg Antibody Per Reaction' (2 Labellings)
Catalog # AATB1294
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    2 Tests
  • Description:
    ReadiLink™ rapid Cy7 antibody labeling kit 'Microscale optimized for labeling 50 µg antibody per reaction'

Specifications

About this item

Cy7 is one of the most popular fluorescent labelling dyes for preparing orange-red fluorescent bioconjugates.

  • Contains all essential components
  • Minimal hands on time
  • Ready to use
  • Robust and simple antibody labelling protocol
  • No column purification required
Caution: For research use only.