ReadiLink™ Rapid Cy3 Antibody Labelling Kit 'Microscale Optimised for Labelling 50 µg Antibody Per Reaction' (2 Labellings)
Katalognr. AATB1290
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:2 Tests
- Description:ReadiLink™ rapid Cy3 antibody labeling kit 'Microscale optimized for labeling 50 µg antibody per reaction'
Specifications
About this item
Cy3 is one of the most popular fluorescent labelling dyes for preparing orange-red fluorescent bioconjugates.
- Contains all essential components
- Minimal hands on time
- Ready to use
- Robust and simple antibody labelling protocol
- No column purification required
Caution: For research use only.