Order Entry
Switzerland
ContactUsLinkComponent
 
ROX Reference Dye *50X Fluorescence Reference Solution for PCR Reactions*
Catalog # AATB400
Supplier:  AAT Bioquest
CAS Number:  
undefined
ROX Reference Dye *50X Fluorescence Reference Solution for PCR Reactions*
Catalog # AATB400
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Description:
    ROX Reference Dye 50X fluorescence reference solution for PCR reactions
  • Purity:
    95%

Specifications

About this item

ROX Reference Dye is equivalent to 12223-012 (Life Technology).

  • Multiple Bioconjugation Applications
  • 95% Purity
Caution: For research use only.