Lumiwox™ Acridinium NHS Ester
Catalog # AATB26000
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:1 mg
- Description:Lumiwox™ acridinium NHS ester
Specifications
About this item
The acridinium NHS ester can be used to label proteins and nucleic acids.
- Chemiluminescent labeling
- Improved sensitivity and stronger signal
- Tolerates hydrogen peroxide reaction
- Suitable for various experimental surfaces
- Multiple applications
Caution: For research use only.