ContactUsLinkComponent
Amplite® Fluorimetric Neuraminidase Assay Kit *Blue Fluorescence*
Amplite® Fluorimetric Neuraminidase Assay Kit *Blue Fluorescence*
Catalog # AATB12602
Supplier:  AAT Bioquest
CAS Number:  
undefined
Amplite® Fluorimetric Neuraminidase Assay Kit *Blue Fluorescence*
Catalog # AATB12602
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • Pk:
    200 Tests
  • Description:
    Amplite™ Fluorimetric neuraminidase assay kit *blue fluorescence*
  • Tests per kit:
    200 Tests
Specifications
About this item
Neuraminidases, also called sialidases, are glycoside hydrolase enzymes that catalyze the hydrolysis of terminal sialic acid residues and neuraminic acid.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.