ContactUsLinkComponent
Amplite® Colorimetric Hydrogen Peroxide Assay Kit
Amplite® Colorimetric Hydrogen Peroxide Assay Kit
Catalog # AATB11500
Supplier:  AAT Bioquest
CAS Number:  
undefined
Amplite® Colorimetric Hydrogen Peroxide Assay Kit
Catalog # AATB11500
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • Pk:
    500 Tests
  • Description:
    Amplite™ colorimetric hydrogen peroxide assay kit
  • Tests per kit:
    500 Tests
Specifications
About this item
Hydrogen peroxide (H2O2) is a reactive oxygen metabolic by-product that serves as a key regulator for a number of oxidative stress-related states.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.