Order Entry
Switzerland
ContactUsLinkComponent
Cell Meter™ Caspase 8 Activity Apoptosis Assay Kit *Red Fluorescence*
Cell Meter™ Caspase 8 Activity Apoptosis Assay Kit *Red Fluorescence*
Catalog # AATB22816
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cell Meter™ Caspase 8 Activity Apoptosis Assay Kit *Red Fluorescence*
Catalog # AATB22816
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    100 Tests
  • Description:
    Cell Meter™ Caspase 8 activity apoptosis assay kit *Red fluorescence*
  • Tests per kit:
    100 Tests

Specifications

About this item

Our Cell Meter™ assay kits are a set of tools for monitoring cellular functions.

  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.