ContactUsLinkComponent
Spexyte™ Intracellular pH Calibration Buffer Kit
Spexyte™ Intracellular pH Calibration Buffer Kit
Catalog # AATB21235
Supplier:  AAT Bioquest
CAS Number:  
undefined
Spexyte™ Intracellular pH Calibration Buffer Kit
Catalog # AATB21235
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • Pk:
    100 Tests
  • Description:
    Spexyte™ intracellular pH calibration buffer kit
  • Tests per kit:
    100 Tests
Specifications
About this item
Intracellular pH (pHi) plays an important modulating role in many cellular events, including cell volume regulation, cellular metabolism, calcium regulation, receptor-mediated signal transduction, ion transport, endocytosis, and other cellular processes.
  • Suitable for all major pH indicators
  • Sensitive and quick results
  • Includes a range of calibration buffers
Caution: For research use only.