ContactUsLinkComponent
Cell Meter™ Fluorometric Intracellular Nitric Oxide (NO) Activity Assay Kit *NIR Fluorescence Optimized For Microplate Reader*
Cell Meter™ Fluorometric Intracellular Nitric Oxide (NO) Activity Assay Kit *NIR Fluorescence Optimized For Microplate Reader*
Catalog # AATB16359
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Cell Meter™ Fluorometric Intracellular Nitric Oxide (NO) Activity Assay Kit *NIR Fluorescence Optimized For Microplate Reader*
Catalog # AATB16359
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    200 Tests
  • Description:
    Cell Meter™ Fluorimetric intracellular Nitric Oxide (NO) activity assay kit *NIR fluorescence optimized for microplate reader*
  • Tests per kit:
    200 Tests
Specifications
About this item
Nitric oxide (NO) is an important biological regulator involved in numbers of physiological and pathological processes.
  • High sensitivity
  • Bright fluorescence
  • Suitable for live cell imaging
Caution: For research use only.