Order Entry
Switzerland
Orders LinkContactUsLinkComponent
LysoBrite™ Orange

LysoBrite™ Orange

LysoBrite™ Orange, 500 Tests, 500 Tests
Catalog # AATB22644
Supplier:   AAT Bioquest
LysoBrite™ Orange
Catalog # AATB22644
Supplier:  AAT Bioquest
CAS Number:  
LysoBrite™ Orange
LysoBrite™ Orange

LysoBrite™ Orange, 500 Tests, 500 Tests

CAS Number:  
Price
SFr. 237.00
Log in for availability
1 * 500 Tests
Product Details & Documents

Lysosomes are cellular organelles which contain acid hydrolase enzymes to break up waste materials and cellular debris.

  • Lysosome-Specific Dye
  • Long-Term Use
  • Low Cytotoxicity

Caution: For research use only.

Specifications

Specifications

  • Pk:
    500 Tests
  • Description:
    LysoBrite™ Orange
  • Tests per kit:
    500 Tests
  • Cat. No.:
    AATB22644
  • Purity:
    95%

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.