DiYO™-1 [equivalent to YOYO®-1] *5 mM DMSO Solution* (0.2 ml)
Catalog # AATB17580
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pk:0,2 mL
- Description:DiYO™-1
- Purity:95%
Specifications
About this item
DiYO™-1 is chemically equivalent to YOYO®-1 (YOYO® is the trademark of Invitrogen).
- Cell-Impermeable
- Useful For Multicolor Imaging
- Bright Fluorescence
5 mM DMSO Solution
Caution: For research use only.