Order Entry
Switzerland
ContactUsLinkComponent
DiYO™-1 [Equivalent To YOYO®-1]
DiYO™-1 [Equivalent To YOYO®-1]
Catalog # AATB17579
Supplier:  AAT Bioquest
CAS Number:  
undefined
DiYO™-1 [Equivalent To YOYO®-1]
Catalog # AATB17579
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Description:
    DiYO™-1
  • Purity:
    95%

Specifications

About this item

DiYO™-1 is chemically equivalent to YOYO®-1 (YOYO® is the trademark of Invitrogen).

  • Cell-Impermeable
  • Useful For Multicolor Imaging
  • Bright Fluorescence
Caution: For research use only.