Order Entry
Switzerland
ContactUsLinkComponent
DL-α-Tocopherol (Vitamin E) 96.0-102.0%
Catalog # SPCMYY2750-1L
Supplier:  Spectrum Chemical
CAS Number:  
DL-α-Tocopherol (Vitamin E) 96.0-102.0%
Catalog # SPCMYY2750-1L
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, amber
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Acidity:
    1.0 ml
  • Purity:
    96 - 102%
  • Grade:
    USP

Specification Test Results

About this item

 :
 :430.71 g/mol
CAS Number:10191-41-0
 :1406-66-2
Formula:C₂₉H₅₀O₂
MDL Number:MFCD00072051