Order Entry
Switzerland
ContactUsLinkComponent
DBCO-Cy5 fluorescent probe
Irritant
DBCO-Cy5 fluorescent probe
Catalog # AATB923
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
DBCO-Cy5 fluorescent probe
Catalog # AATB923
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
ADR:5,II
 :1211.6 g/mol
 :GHS07
CAS Number:1564286-24-3