Skip to Header
Skip to Main Content
Skip to Footer
Erweiterte Suche
Erweiterte Suche
Sicherheitsdatenblatt (SDS)
Zertifikate
Herstellerdaten
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1X4 Color™ V405 Dye Set
1X4 Color™ V405 Dye Set
1X4 Color™ V405 Dye Set
Reizend
1X4 Color™ V405 Dye Set
Catalog # AATB8000
Anbieter:
AAT Bioquest
CAS-Nummer:
Expand image
Reizend
1X4 Color™ V405 Dye Set
Catalog # AATB8000
Anbieter:
AAT Bioquest
CAS-Nummer:
Technische Daten
Pack type:
Plastic bottle
Pk:
1 SET
Seq:
0125
Technische Daten
Über diesen Artikel
Produktdetails
:
ADR:
5,II
:
GHS07
Produktdetails