576/589 NHS Ester, Bodi Fluor™ fluorescent probe
Plastic bottle, 5 mg
Catalog # AATB661
Supplier: AAT Bioquest
576/589 NHS Ester, Bodi Fluor™ fluorescent probe
Catalog # AATB661
Supplier: AAT Bioquest
CAS Number: 201998-61-0
Jump to:
Product Details & Documents
Bodi Fluor™ 576/589 is chemically the same molecule to BODIPY™ 576/589 dye.
- Bright signal
- Highly cell permeable
- High extinction coefficient
- Insensitive to polarity/pH
- High fluorescence quantum yield
Synonyms: Py-BODIPY-NHS ester, 2,5-Dioxopyrrolidin-1-yl 3-(5,5-difluoro-7-(1H-pyrrol-2-yl)-5H-5l4,6l4-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinin-3-yl)propanoate
- CAS Number:201998-61-0
- MDL Number:MFCD32858277
- Molecular Weight:426.19 g/mol
- ADR:5,II
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:5 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



