Biotin-dT Phosphoramidite
Supplier: AAT Bioquest
Certificates
About this item
CAS Number:198080-40-9
:
Formula:C₆₉H₈₉N₈O₁₂PS
:1285.55 g/mol
Frequently Bought Together
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
Serological pipettes, Stripette™
Supplier: Corning
Multiwell cell culture plates, Falcon®
Supplier: Corning
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Individually Wrapped Serological Pipettes, Falcon®
Supplier: Corning
VWR®, Microcentrifuge Tubes with Socket Screw Caps
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom, 5 ml
Supplier: VWR Collection
VWR®, Petri Dish, PS
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
J.T.Baker®, Plates, Square Well
Supplier: Avantor
VWR® Water Sampling Bottles, with Tamper Evident Cap
Supplier: VWR Collection
Centrifuge tubes, PP, with screw cap
Supplier: KARTELL
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
VWR®, Transfer Pipettes, Graduated
Supplier: VWR Collection
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
Serological pipettes
Supplier: Greiner Bio-One
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF