Order Entry
Switzerland
ContactUsLinkComponent
7-Hydroxy-4-methylcoumarin-3-acetic acid succinimidyl ester
7-Hydroxy-4-methylcoumarin-3-acetic acid succinimidyl ester
Catalog # AATB556
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
7-Hydroxy-4-methylcoumarin-3-acetic acid succinimidyl ester
Catalog # AATB556
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :331.28 g/mol
CAS Number:96735-88-5
Formula:C₁₆H₁₃NO₇
MDL Number:MFCD00467590