SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
FMOC-Glu(5-TAMRA)-OH
FMOC-Glu(5-TAMRA)-OH for biomolecular labeling
FMOC-Glu(5-TAMRA)-OH for biomolecular labeling
AATB5015
:
AAT Bioquest
:
FMOC-Glu(5-TAMRA)-OH for biomolecular labeling
AATB5015
:
AAT Bioquest
:
Pack type:
Plastic bottle
Pk:
100 mg
Seq:
2440
:
:
5,II
:
GHS07