Order Entry
Switzerland
ContactUsLinkComponent
 
6-ROX azide ≥90%
Catalog # AATB494
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
6-ROX azide ≥90%
Catalog # AATB494
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :602.68 g/mol