Order Entry
Switzerland
ContactUsLinkComponent
Biotinyl tyramide for signal amplification
Irritant
Biotinyl tyramide for signal amplification
Catalog # AATB45350
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
Biotinyl tyramide for signal amplification
Catalog # AATB45350
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :363.5 g/mol
ADR:5,II
 :GHS07
CAS Number:41994-02-9
Formula:C₁₈H₂₅N₃O₃S
MDL Number:MFCD27953026