Springen zu Kopfbereich
Springen zu Hauptinhalt
Springen zu Fußzeile
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
iFluor®
iFluor® 555, Styramide *Superior Replacement for Alexa Fluor 555 tyramide*
iFluor® 555, Styramide *Superior Replacement for Alexa Fluor 555 tyramide*
iFluor® 555, Styramide *Superior Replacement for Alexa Fluor 555 tyramide*
Catalog # AATB45027
Supplier:
AAT Bioquest
CAS Number:
Expand image
iFluor® 555, Styramide *Superior Replacement for Alexa Fluor 555 tyramide*
Catalog # AATB45027
Supplier:
AAT Bioquest
CAS Number:
Specifications
Pk:
100
Seq:
0100
Specifications
About this item
Produktdetails
:
Produktdetails