SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Cy5 biotin conjugate
Cy5 biotin conjugate biodetection
Cy5 biotin conjugate biodetection
Irritant
Cy5 biotin conjugate biodetection
Catalog # AATB3100
Supplier:
AAT Bioquest
CAS Number:
Expand image
Irritant
Cy5 biotin conjugate biodetection
Catalog # AATB3100
Supplier:
AAT Bioquest
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
5 mg
Seq:
2210
Specifications
About this item
Product Details
:
:
1353.82 g/mol
ADR:
5,II
:
GHS07
Product Details