Order Entry
Switzerland
ContactUsLinkComponent
 
5-Carboxy-4,7-dichlorortetramethylrhodamine (5-dTMR) ≥95%
  AATB305
 :  
undefined
5-Carboxy-4,7-dichlorortetramethylrhodamine (5-dTMR) ≥95%
  AATB305
 :  

 

  • Pk:
    25 mg
  • Seq:
    2315

 

 

 :
 :600.54 g/mol