Order Entry
Switzerland
ContactUsLinkComponent
D-Biotin-N-succinimidyl ester ≥90%
Irritant
D-Biotin-N-succinimidyl ester ≥90%
Catalog # AATB3002
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
D-Biotin-N-succinimidyl ester ≥90%
Catalog # AATB3002
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :341.38 g/mol
ADR:5,II
 :GHS07
CAS Number:35013-72-0
Formula:C₁₄H₁₉N₃O₅S
MDL Number:MFCD00078531