
PE-iFluor™ 594 Tandem, ReadiUse™ fluorescent probe, preactivated
Catalog # AATB2584
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 mg
- Seq:2070
Specifications
About this item
:
ADR:5,II
:240000 g/mol
:GHS07