Cy2DIGE NHS ester ≥90%
100 Nmo
Catalog # AATB25300
Supplier: AAT Bioquest
Jump to:
Product Details & Documents
Cy2DIGE NHS ester, an equivalent of Cy2® NHS ester minimal dye, is used for labeling proteins subject to DIGE analysis.
- Sensitive Detection
- Bright Fluorescence
Caution: For research use only.
- Molecular Weight:677.5 g/mol
Specifications
Specifications
- Pk:100 Nmo