Order Entry
Switzerland
ContactUsLinkComponent
DASPEI (2-(4-(Dimethylamino)styryl)-N-ethylpyridinium iodide) ≥95%
Irritant
DASPEI (2-(4-(Dimethylamino)styryl)-N-ethylpyridinium iodide) ≥95%
Catalog # AATB22225
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
DASPEI (2-(4-(Dimethylamino)styryl)-N-ethylpyridinium iodide) ≥95%
Catalog # AATB22225
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :380.273 g/mol
ADR:5,II
 :GHS07
CAS Number:3785-01-1
Formula:C₁₇H₂₁IN₂
MDL Number:MFCD00011994