Order Entry
Switzerland
ContactUsLinkComponent
1,1'-Di-n-octadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate ≥95%
1,1'-Di-n-octadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate ≥95%
Catalog # AATB22102
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
1,1'-Di-n-octadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate ≥95%
Catalog # AATB22102
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :933.885 g/mol
ADR:5,II
 :GHS07
CAS Number:41085-99-8
Formula:C₅₉H₉₇ClN₂O₄
MDL Number:MFCD00142354