Order Entry
Switzerland
ContactUsLinkComponent
 
3,3'-Diethylthiatricarbocyanine iodide ≥95%
Catalog # AATB22077
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
3,3'-Diethylthiatricarbocyanine iodide ≥95%
Catalog # AATB22077
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :544.52 g/mol
ADR:5,II
 :GHS07
CAS Number:3071-70-3
Formula:C₂₅H₂₅IN₂S₂
MDL Number:MFCD00041994