Order Entry
Switzerland
ContactUsLinkComponent
 
3,3'-Diethylthiacarbocyanine iodide [DiSC2(3)] ≥95%
Catalog # AATB22073
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
3,3'-Diethylthiacarbocyanine iodide [DiSC2(3)] ≥95%
Catalog # AATB22073
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :492.44 g/mol
CAS Number:905-97-5
Formula:C₂₁H₂₁IN₂S₂
MDL Number:MFCD00011956