1,1'-Dioctadecyl-3,3,3',3'-tetramethylindodicarbocyanine (DID) 5 mM in DMSO ≥95% for labeling
Catalog # AATB22033
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:10 mL
- Seq:2275
Specifications
About this item
:
:959.923 g/mol
CAS Number:127274-91-3
Formula:C₆₁H₉₉ClN₂O₄