Order Entry
Switzerland
ContactUsLinkComponent
Tide Quencher™ 1 CPG 500 Å, Tide Quencher™ fluorescent probe
Irritant
Tide Quencher™ 1 CPG 500 Å, Tide Quencher™ fluorescent probe
Catalog # AATB2193
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
Tide Quencher™ 1 CPG 500 Å, Tide Quencher™ fluorescent probe
Catalog # AATB2193
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
ADR:5,II
 :GHS07