Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dextran
Dextran, M.W. 150,000 TRITC-labeled
Dextran, M.W. 150,000 TRITC-labeled
Dextran, M.W. 150,000 TRITC-labeled
Catalog # AATB21710
Supplier:
AAT Bioquest
CAS Number:
Expand image
Dextran, M.W. 150,000 TRITC-labeled
Catalog # AATB21710
Supplier:
AAT Bioquest
CAS Number:
Specifications
Pack type:
Vial
Pk:
25 mg
Seq:
2315
Specifications
About this item
Product Details
:
CAS Number:
9004-54-0
Formula:
(C₆H₁₀O₅)n
MDL Number:
MFCD00130935
Product Details