Springen zu Kopfbereich
Springen zu Hauptinhalt
Springen zu Fußzeile
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
MEQ
MEQ chloride indicator
MEQ chloride indicator
MEQ chloride indicator
AATB21250
:
AAT Bioquest
:
MEQ chloride indicator
AATB21250
:
AAT Bioquest
:
Pack type:
Plastic bottle
Pk:
100 mg
Seq:
2440
:
:
315.15 g/mol
:
5,II
:
GHS07
:
34373-76-7