Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tide Quencher™ 7WS alkyne
Tide Quencher™ 7WS alkyne, Tide Quencher™ fluorescent probe
Tide Quencher™ 7WS alkyne, Tide Quencher™ fluorescent probe
AATB2113
:
AAT Bioquest
:
Tide Quencher™ 7WS alkyne, Tide Quencher™ fluorescent probe
AATB2113
:
AAT Bioquest
:
Pack type:
Plastic bottle
Pk:
1 mg
Seq:
2070
:
:
953.3 g/mol
:
5,II
:
GHS07