
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) ≥95%
Supplier: AAT BIOQUEST
Certificates
About this item
CAS Number:28718-90-3
:
Formula:C₁₆H₁₅N₅·2HCl
MDL Number:MFCD00012681
:350.25 g/mol
ADR:5,II
:GHS07
Frequently Bought Together