Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Express Eingabe
Bestellstatus
Ihr Konto
Warenkorb
Switzerland
Express Eingabe
Bestellstatus
Ihr Konto
Warenkorb
ContactUsLinkComponent
Heim
Cyanine-3- dUTP [Cy3-dUTP]
Cyanine-3- dUTP [Cy3-dUTP] 1 mM in TRIS buffer (pH 7.5)
Cyanine-3- dUTP [Cy3-dUTP] 1 mM in TRIS buffer (pH 7.5)
Irritant
Cyanine-3- dUTP [Cy3-dUTP] 1 mM in TRIS buffer (pH 7.5)
Catalog # AATB17025
Supplier:
AAT Bioquest
CAS Number:
Expand image
Irritant
Cyanine-3- dUTP [Cy3-dUTP] 1 mM in TRIS buffer (pH 7.5)
Catalog # AATB17025
Supplier:
AAT Bioquest
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
25 Nmo
Seq:
0311
Specifications
About this item
Product Details
:
:
1150 g/mol
ADR:
5,II
:
GHS07
Product Details