Order Entry
Switzerland
ContactUsLinkComponent
TF3-dUTP 1 mM in TRIS buffer (pH 7.5)
Irritant
TF3-dUTP 1 mM in TRIS buffer (pH 7.5)
Catalog # AATB17008
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
TF3-dUTP 1 mM in TRIS buffer (pH 7.5)
Catalog # AATB17008
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 Nmo
  • Seq:
    0311

Specifications

About this item

 :
 :1150 g/mol
ADR:5,II
 :GHS07