Skip to Header
Skip to Main Content
Skip to Footer
Erweiterte Suche
Erweiterte Suche
Sicherheitsdatenblatt (SDS)
Zertifikate
Herstellerdaten
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2',7'-Dichlorofluorescin diacetate
2',7'-Dichlorofluorescin diacetate ≥95%
2',7'-Dichlorofluorescin diacetate ≥95%
AATB15204
:
AAT Bioquest
:
2',7'-Dichlorofluorescin diacetate ≥95%
AATB15204
:
AAT Bioquest
:
Pack type:
Plastic bottle
Pk:
25 mg
Seq:
2315
:
:
487.285 g/mol
:
5,II
:
GHS07
:
4091-99-0
:
C₂₄H₁₆Cl₂O₇
:
MFCD00128955