Order Entry
Switzerland
ContactUsLinkComponent
Cyanine 3-NHS ester chloride ≥90% [equivalent to Cy3® NHS ester]
Cyanine 3-NHS ester chloride ≥90% [equivalent to Cy3® NHS ester]
Catalog # AATB141
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Cyanine 3-NHS ester chloride ≥90% [equivalent to Cy3® NHS ester]
Catalog # AATB141
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :590.15 g/mol
CAS Number:146368-16-3
Formula:C₃₄H₄₀ClN₃O₄