Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
phosphatase reaction stopping solution
phosphatase reaction stopping solution, Signal Guard™
phosphatase reaction stopping solution, Signal Guard™
Irritant
phosphatase reaction stopping solution, Signal Guard™
Catalog # AATB11622
Supplier:
AAT Bioquest
CAS Number:
Expand image
Irritant
phosphatase reaction stopping solution, Signal Guard™
Catalog # AATB11622
Supplier:
AAT Bioquest
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
100 mL
Seq:
2440
Specifications
About this item
Product Details
:
ADR:
5,II
:
GHS07
Product Details