Fluorescein ≥95%
Catalog # AATB1
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:332.31 g/mol
ADR:5,II
:GHS07
CAS Number:2321-07-5
Formula:C₂₀H₁₂O₅
MDL Number:MFCD00005050