Orders LinkContactUsLinkComponent
Spare burettes, Dr. Schilling Pattern
Spare burettes, Dr. Schilling Pattern
Supplier:   Brand
Spare burettes, Dr. Schilling Pattern
Supplier:  Brand
Certificates
Spare burettes, Dr. Schilling Pattern
Spare burettes, Dr. Schilling Pattern
6 Options Available Below
Product Details & Documents
AR® glass, class B.
  • Calibrated to deliver 'Ex'
  • Schellbach stripe, reduced distance between subdivision marks
DIN EN ISO 385
Bottle requirements: 500 ml for 10 and 15 ml; 1000 ml for 25 and 50 ml
Certifications: DIN EN ISO 385.
Product Family Options
Clear glass
Product Information
  • Capacity
    Length
    Availability
    Price
  • 10 ml
    340 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 61.20
    SFr. 61.20
    1 * 1 ST
       SFr. 61.20
  • 15 ml
    320 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 61.20
    SFr. 61.20
    1 * 1 ST
       SFr. 61.20
  • 25 ml
    390 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 64.60
    SFr. 64.60
    1 * 1 ST
       SFr. 64.60
  • 50 ml
    600 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 64.60
    SFr. 64.60
    1 * 1 ST
       SFr. 64.60
Amber glass
Product Information
  • Capacity
    Length
    Availability
    Price
  • 25 ml
    420 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 82.60
    SFr. 82.60
    1 * 1 ST
       SFr. 82.60
  • 50 ml
    670 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 82.60
    SFr. 82.60
    1 * 1 ST
       SFr. 82.60