Vitamin A palmitate in water, colourless
Catalog # CAMBCLM-10838-PK
Supplier: CAMBRIDGE ISOTOPE LABS
CAS Number:
Specifications
- Pk:0,5 g
- Seq:3035
Specifications
About this item
:
:524.87 g/mol
Melting Pt:100 °C (212 °F)
Flash Pt:194 °C (381 °F)
:GHS08
CAS Number:79-81-2
Formula:C₃₆H₆₀O₂
MDL Number:MFCD00019414